COMH4211B8B6 Combi-Blocks, Inc.

2-(2-Bromoethoxy)-4-fluoro-1-methoxybenzene

  • MDL number MFCD31613703
  • Cas Number : 125960-38-5
  • Molecular Weight : 249.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5439

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5439
Availability details for
Quantity
  • Availability
  • Add to Cart