COMH4211B8B7 Combi-Blocks, Inc.

Niranthin

  • MDL number MFCD09970365
  • Cas Number : 50656-77-4
  • Molecular Weight : 432.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5443

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5443
Availability details for
Quantity
  • Availability
  • Add to Cart