COMH4211B8B9 Combi-Blocks, Inc.

Ethyl 2-methyl-4-vinylbenzoate

  • MDL number MFCD31613790
  • Cas Number : 1220869-25-9
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5446

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5446
Availability details for
Quantity
  • Availability
  • Add to Cart