COMH4211B8BC Combi-Blocks, Inc.

Benzonitrile, 4-amino-2-(phenylmethoxy)-

  • MDL number MFCD13192719
  • Cas Number : 284044-40-2
  • Molecular Weight : 224.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-amino-2-(benzyloxy)benzonitrile
Inchi Code 1S/C14H12N2O/c15-9-12-6-7-13(16)8-14(12)17-10-11-4-2-1-3-5-11/h1-8H,10,16H2
Inchi Key BACKXVWDHDEIHG-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5457

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5457
Availability details for
Quantity
  • Availability
  • Add to Cart