COMH4211B8BD Combi-Blocks, Inc.

Trans-4-(methoxymethyl)cyclohexanamine hydrochloride

  • MDL number MFCD28383638
  • Cas Number : 919799-81-8
  • Molecular Weight : 179.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1r,4r)-4-(methoxymethyl)cyclohexan-1-amine hydrochloride
Inchi Code 1S/C8H17NO.ClH/c1-10-6-7-2-4-8(9)5-3-7;/h7-8H,2-6,9H2,1H3;1H/t7-,8-;
Inchi Key BDNBGXGBMVAZBM-KMMPGQJCSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5458

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5458
Availability details for
Quantity
  • Availability
  • Add to Cart