COMH4211B8BF Combi-Blocks, Inc.

3-Bromo-5-isobutyrylamino-benzoic acid

  • MDL number MFCD24125691
  • Cas Number : 1499086-15-5
  • Molecular Weight : 286.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5462

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5462
Availability details for
Quantity
  • Availability
  • Add to Cart