COMH4211B8C0 Combi-Blocks, Inc.

1-(2-Fluoro-4-trifluoromethyl-phenyl)-propylamine hydrochloride

  • MDL number MFCD31539567
  • Cas Number : 2203940-65-0
  • Molecular Weight : 257.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5463

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5463
Availability details for
Quantity
  • Availability
  • Add to Cart