COMH4211B8C3 Combi-Blocks, Inc.

Cyclobutylmethyl-(4-fluoro-phenyl)-amine

  • MDL number MFCD03211331
  • Cas Number : 356539-70-3
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5468

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5468
Availability details for
Quantity
  • Availability
  • Add to Cart