COMH4211B8C4 Combi-Blocks, Inc.

2-Amino-4,5-difluoro-n,n-dimethylbenzamide

  • MDL number MFCD16159064
  • Cas Number : 1248034-89-0
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5470

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5470
Availability details for
Quantity
  • Availability
  • Add to Cart