COMH4211B8C9 Combi-Blocks, Inc.

5-Bromo-2-(4-methoxy-benzylamino)-nicotinic acid tert-butyl ester

  • MDL number MFCD31557910
  • Cas Number : 2206609-88-1
  • Molecular Weight : 393.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5478

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5478
Availability details for
Quantity
  • Availability
  • Add to Cart