COMH4211B917 Combi-Blocks, Inc.

2-(2-Methoxyethoxy)-5-methylaniline

  • MDL number MFCD08688035
  • Cas Number : 156564-83-9
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5481

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5481
Availability details for
Quantity
  • Availability
  • Add to Cart