COMH4211B918 Combi-Blocks, Inc.

3'-Methyl-4'-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-biphenyl-3-carboxylic acid methyl ester

  • MDL number MFCD31579464
  • Molecular Weight : 352.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5483

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5483
Availability details for
Quantity
  • Availability
  • Add to Cart