COMH4211B919 Combi-Blocks, Inc.

1-(2-Methylpropyl)piperidine-4-carbonitrile

  • MDL number MFCD11934558
  • Cas Number : 1181456-18-7
  • Molecular Weight : 166.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5487

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5487
Availability details for
Quantity
  • Availability
  • Add to Cart