COMH4211B91A Combi-Blocks, Inc.

2-Bromo-1-(cyclopropylmethoxy)-4-nitrobenzene

  • MDL number MFCD29095257
  • Cas Number : 1369836-75-8
  • Molecular Weight : 272.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5490

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5490
Availability details for
Quantity
  • Availability
  • Add to Cart