COMH4211B91B Combi-Blocks, Inc.

N-[(4-Methyl-3-nitrophenyl)methyl]cyclopropanamine

  • MDL number MFCD12146403
  • Cas Number : 1152957-06-6
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5494

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5494
Availability details for
Quantity
  • Availability
  • Add to Cart