COMH4211B91C Combi-Blocks, Inc.

6-Iodo-8-trifluoromethyl-imidazo[1,2-a]pyridine-2-carboxylic acid methyl ester

  • MDL number MFCD31381598
  • Cas Number : 1237841-44-9
  • Molecular Weight : 370.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5498

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5498
Availability details for
Quantity
  • Availability
  • Add to Cart