COMH4211B91D Combi-Blocks, Inc.

4-Bromo-2-(4-methoxyphenyl)thiazole

  • MDL number MFCD11869497
  • Cas Number : 1078734-03-8
  • Molecular Weight : 270.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(4-bromo-1,3-thiazol-2-yl)phenyl methyl ether
Inchi Code 1S/C10H8BrNOS/c1-13-8-4-2-7(3-5-8)10-12-9(11)6-14-10/h2-6H,1H3
Inchi Key MUNWKDJOWYMCTP-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5500

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5500
Availability details for
Quantity
  • Availability
  • Add to Cart