COMH4211B91E Combi-Blocks, Inc.

4-Acetyl-3-chlorobenzoic acid

  • MDL number MFCD20644295
  • Cas Number : 57542-72-0
  • Molecular Weight : 198.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5503

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5503
Availability details for
Quantity
  • Availability
  • Add to Cart