COMH4211B91F Combi-Blocks, Inc.

1-Bromo-3-(2-ethoxy-ethyl)-benzene

  • MDL number MFCD31628728
  • Cas Number : 2270910-83-1
  • Molecular Weight : 229.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5504

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5504
Availability details for
Quantity
  • Availability
  • Add to Cart