COMH4211B920 Combi-Blocks, Inc.

3-(4,4-Difluoropiperidin-1-yl)-2,2-dimethylpropionic acid

  • MDL number MFCD30486330
  • Cas Number : 1889794-96-0
  • Molecular Weight : 221.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5508

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5508
Availability details for
Quantity
  • Availability
  • Add to Cart