COMH4211B921 Combi-Blocks, Inc.

2-Chloro-n-(3-nitrophenyl)pyridine-4-amine

  • MDL number MFCD28672734
  • Cas Number : 1268619-67-5
  • Molecular Weight : 249.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5509

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5509
Availability details for
Quantity
  • Availability
  • Add to Cart