COMH4211B922 Combi-Blocks, Inc.

3-(2-Ethoxy-3-fluoro-phenyl)-propionic acid

  • MDL number MFCD28847782
  • Cas Number : 1804283-67-7
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5510

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5510
Availability details for
Quantity
  • Availability
  • Add to Cart