COMH4211B924 Combi-Blocks, Inc.

(4-Amino-2-chlorophenyl)(4-methylpiperazin-1-yl)methanone

  • MDL number MFCD09050352
  • Cas Number : 919116-51-1
  • Molecular Weight : 253.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5514

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5514
Availability details for
Quantity
  • Availability
  • Add to Cart