COMH4211B926 Combi-Blocks, Inc.

1-(4-(Benzyloxy)-5-chloro-2-hydroxyphenyl)ethanone

  • MDL number MFCD22479609
  • Cas Number : 1822806-82-5
  • Molecular Weight : 276.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5527

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5527
Availability details for
Quantity
  • Availability
  • Add to Cart