COMH4211B928 Combi-Blocks, Inc.

5-Chloro-n-(propan-2-yl)pyrazine-2-carboxamide

  • MDL number MFCD29959549
  • Cas Number : 1637677-38-3
  • Molecular Weight : 199.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5532

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5532
Availability details for
Quantity
  • Availability
  • Add to Cart