COMH4211B929 Combi-Blocks, Inc.

(2-Fluoro-4-methyl-phenyl)-piperazin-1-yl-methanone

  • MDL number MFCD22932052
  • Cas Number : 1525482-27-2
  • Molecular Weight : 222.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5533

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5533
Availability details for
Quantity
  • Availability
  • Add to Cart