COMH4211B92A Combi-Blocks, Inc.

tert-Butyl 2-(4-hydroxy-1h-pyrazol-1-yl)acetate

  • MDL number MFCD25968386
  • Cas Number : 883976-91-8
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5534

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5534
Availability details for
Quantity
  • Availability
  • Add to Cart