COMH4211B92B Combi-Blocks, Inc.

Piperidin-1-yl-[4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyrazol-1-yl]-methanone

  • MDL number MFCD31583376
  • Molecular Weight : 305.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name piperidin-1-yl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)methanone
Inchi Code 1S/C15H24BN3O3/c1-14(2)15(3,4)22-16(21-14)12-10-17-19(11-12)13(20)18-8-6-5-7-9-18/h10-11H,5-9H2,1-4H3
Inchi Key GQDVSZZOPQGPDH-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5535

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5535
Availability details for
Quantity
  • Availability
  • Add to Cart