COMH4211B92D Combi-Blocks, Inc.

1-(2-Cyclopropylmethoxy-3-fluoro-phenyl)-ethylamine hydrochloride

  • MDL number MFCD31557792
  • Cas Number : 2206608-27-5
  • Molecular Weight : 245.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5543

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5543
Availability details for
Quantity
  • Availability
  • Add to Cart