COMH4211B92F Combi-Blocks, Inc.

5-Fluoro-2-(2-methoxyethoxy)aniline

  • MDL number MFCD08687838
  • Cas Number : 946683-70-1
  • Molecular Weight : 185.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5549

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5549
Availability details for
Quantity
  • Availability
  • Add to Cart