COMH4211B932 Combi-Blocks, Inc.

2-(4-Chlorophenyl)-2,2-difluoroethanol

  • MDL number MFCD28038847
  • Cas Number : 207799-86-8
  • Molecular Weight : 192.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5553

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5553
Availability details for
Quantity
  • Availability
  • Add to Cart