COMH4211B933 Combi-Blocks, Inc.

Methyl 5-(4-bromo-3-fluorophenoxy)-2-nitrobenzoate

  • MDL number MFCD31614107
  • Cas Number : 1192319-50-8
  • Molecular Weight : 370.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5554

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5554
Availability details for
Quantity
  • Availability
  • Add to Cart