COMH4211B935 Combi-Blocks, Inc.

3-Amino-6-bromopyrazine-2-carbohydrazide

  • MDL number MFCD11045628
  • Cas Number : 1225062-23-6
  • Molecular Weight : 232.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5557

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5557
Availability details for
Quantity
  • Availability
  • Add to Cart