COMH4211B938 Combi-Blocks, Inc.

2-Fluoro-4-piperazin-1-yl-benzoic acid benzyl ester dihydrochloride

  • MDL number MFCD31579529
  • Cas Number : 2270911-82-3
  • Molecular Weight : 387.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5563

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5563
Availability details for
Quantity
  • Availability
  • Add to Cart