COMH4211B93A Combi-Blocks, Inc.

2-(4-Bromo-2-fluoro-phenyl)-1-pyrrolidin-1-yl-ethanone

  • MDL number MFCD25954354
  • Cas Number : 1022931-92-5
  • Molecular Weight : 286.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5566

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5566
Availability details for
Quantity
  • Availability
  • Add to Cart