COMH4211B93B Combi-Blocks, Inc.

3-Bromo-4-(2-methoxyethoxy)benzonitrile

  • MDL number MFCD24307527
  • Cas Number : 1557322-67-4
  • Molecular Weight : 256.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5569

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5569
Availability details for
Quantity
  • Availability
  • Add to Cart