COMH4211B93F Combi-Blocks, Inc.

Benzoic acid, 4-bromo-3-(3-methoxypropoxy)-, methyl ester

  • MDL number MFCD26398528
  • Cas Number : 895240-78-5
  • Molecular Weight : 303.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5581

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5581
Availability details for
Quantity
  • Availability
  • Add to Cart