COMH4211B940 Combi-Blocks, Inc.

1-(3-Chloro-5-fluorophenyl)propan-1-amine

  • MDL number MFCD18704117
  • Cas Number : 1270493-39-4
  • Molecular Weight : 187.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5585

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5585
Availability details for
Quantity
  • Availability
  • Add to Cart