COMH4211B942 Combi-Blocks, Inc.

4-(2,2-Difluoroethoxy)-2-fluorobenzonitrile

  • MDL number MFCD23903716
  • Cas Number : 1539535-70-0
  • Molecular Weight : 201.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5588

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5588
Availability details for
Quantity
  • Availability
  • Add to Cart