COMH4211B943 Combi-Blocks, Inc.

5-(3-Methylbutyl)-1h-pyrazole-3-carboxylic acid ethyl ester

  • MDL number MFCD26386003
  • Cas Number : 856256-91-2
  • Molecular Weight : 210.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5589

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5589
Availability details for
Quantity
  • Availability
  • Add to Cart