COMH4211B945 Combi-Blocks, Inc.

5-Bromo-n,n-bis(4-methoxybenzyl)pyridin-3-amine

  • MDL number MFCD31613758
  • Cas Number : 1864768-57-9
  • Molecular Weight : 413.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5596

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5596
Availability details for
Quantity
  • Availability
  • Add to Cart