COMH4211B948 Combi-Blocks, Inc.

2-(Cyclopropylmethoxy)-5-hydroxy-n,n-dimethylbenzamide

  • MDL number MFCD16997978
  • Cas Number : 1243279-57-3
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5603

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5603
Availability details for
Quantity
  • Availability
  • Add to Cart