COMH4211B94C Combi-Blocks, Inc.

5-(1-(Tetrahydro-2h-pyran-2-yl)-1h-pyrazol-4-yl)-1h-indole

  • MDL number MFCD31561616
  • Cas Number : 1808957-23-4
  • Molecular Weight : 267.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5611

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5611
Availability details for
Quantity
  • Availability
  • Add to Cart