COMH4211B94E Combi-Blocks, Inc.

1-(3-Fluoro-2-methylphenyl)ethan-1-amine

  • MDL number MFCD18702592
  • Cas Number : 1270453-63-8
  • Molecular Weight : 153.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5613

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5613
Availability details for
Quantity
  • Availability
  • Add to Cart