COMH4211B950 Combi-Blocks, Inc.

(4-(4-Chloro-3-(trifluoromethyl)phenoxy)phenyl)methanamine

  • MDL number MFCD21364661
  • Cas Number : 1359656-24-8
  • Molecular Weight : 301.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5618

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5618
Availability details for
Quantity
  • Availability
  • Add to Cart