COMH4211B952 Combi-Blocks, Inc.

2-Chloro-4-cyclopropylaniline

  • MDL number MFCD14585512
  • Cas Number : 78242-79-2
  • Molecular Weight : 167.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5623

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5623
Availability details for
Quantity
  • Availability
  • Add to Cart