COMH4211B955 Combi-Blocks, Inc.

(2-Chloro-pyrimidin-5-yloxy)-acetic acid tert-butyl ester

  • MDL number MFCD31618944
  • Cas Number : 1911650-68-4
  • Molecular Weight : 244.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5629

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5629
Availability details for
Quantity
  • Availability
  • Add to Cart