COMH4211B959 Combi-Blocks, Inc.

2-Amino-5-bromo-n,n-dimethylnicotinamide

  • MDL number MFCD16688694
  • Cas Number : 1092580-97-6
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5636

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5636
Availability details for
Quantity
  • Availability
  • Add to Cart