COMH4211B95A Combi-Blocks, Inc.

3-(Tetrahydro-2h-pyran-4-ylmethoxy)benzoic acid

  • MDL number MFCD08669641
  • Cas Number : 1008773-62-3
  • Molecular Weight : 236.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5637

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5637
Availability details for
Quantity
  • Availability
  • Add to Cart