COMH4211B95D Combi-Blocks, Inc.

N-(5-Bromopyridin-3-yl)cyclopropanesulfonamide

  • MDL number MFCD17169876
  • Cas Number : 1083326-19-5
  • Molecular Weight : 277.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5640

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5640
Availability details for
Quantity
  • Availability
  • Add to Cart